Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042655.1 | NP_001036120.1 | 4 | 443 | ||
GenBank mRNA | AK046883.1 | - | 4 | 277 | ||
AK081318.1 | - | 4 | 255 | |||
AK083153.1 | - | 4 | 260 | |||
AK087754.1 | - | 268 | ||||
AK136705.1 | - | 4 | 268 | |||
AK149917.1 | - | 4 | 255 | |||
AK153732.1 | - | 4 | 255 | |||
AK170821.1 | - | 4 | 255 | |||
BC017607.1 | - | 4 | 443 | 4507589 | ||
BC035449.1 | - | 4 | 242 |