Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144946.4 | NP_659195.3 | 3 | 1086 | ||
GenBank mRNA | AF448840.1 | - | 3 | 419 | ||
AK043710.1 | - | 3 | 422 | |||
AK081325.1 | - | 3 | 1087 | |||
AY138990.1 | - | 3 | 155 | |||
BC051145.1 | - | 3 | 446 | 6590345 |