Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145099.2 | NP_659567.2 | 4 | 342 | ||
GenBank mRNA | AB083654.1 | - | 4 | 318 | ||
AF510316.1 | - | 4 | 342 | |||
BC108984.1 | - | 4 | 328 | 40047664 | ||
BC108985.1 | - | 4 | 328 |