Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mchr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145132.2 | NP_660114.1 | 2 | 384 | ||
GenBank mRNA | AF498247.1 | - | 2 | 109 | ||
AF498248.1 | - | 2 | 109 | |||
AK134867.1 | - | 2 | 267 | |||
BC128285.1 | - | 2 | 109 | |||
BC128286.1 | - | 2 | 109 | 40102363 |
Set Membership: |
G-protein coupled receptor |