Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172100.1 | NP_001165571.1 | 5 | 622 | ||
NM_001172101.1 | NP_001165572.1 | 5 | 625 | |||
NM_145135.3 | NP_660117.2 | 5 | 773 | |||
GenBank mRNA | AF071546.1 | - | 4 | 493 | ||
AK133822.1 | - | 5 | 553 | |||
AK141351.1 | - | 5 | 598 | |||
AK145173.1 | - | 5 | 607 | |||
AK147059.1 | - | 5 | 559 | |||
BC010331.1 | - | 5 | 642 | 2654341 |