Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145139.2 | NP_660121.2 | 3 | 183 | ||
GenBank mRNA | AJ310346.1 | - | 3 | 193 | ||
AK032936.1 | - | 3 | 171 | |||
AK075584.1 | - | 3 | 189 | |||
AK150895.1 | - | 3 | 178 | |||
AK151175.1 | - | 3 | 178 | |||
AK167640.1 | - | 3 | 178 | |||
AK168009.1 | - | 3 | 181 | |||
AK168295.1 | - | 3 | 180 | |||
BC024463.1 | - | 3 | 183 | 4975621 |