Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145144.1 | NP_660126.1 | 2, 3 | 137 | ||
GenBank mRNA | AB035322.1 | - | 2, 3 | 89 | ||
AK028955.1 | - | 2, 3 | 95 | |||
AK045539.1 | - | 2, 3 | 167 | |||
BC024599.1 | - | 2, 3 | 137 | 5044495 |