Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000735.3 | NP_000726.1 | 3 | 261 | ||
NM_001252383.1 | NP_001239312.1 | 4 | 354 | |||
GenBank mRNA | AK311972.1 | - | 3 | 218 | ||
BC020782.1 | - | 3 | 220 | 4732686 | ||
BC055080.1 | - | 3 | 222 | 4044086 | ||
BG571584.1 | - | 4 | 311 | |||
BG619154.1 | - | 3 | 262 | |||
CR457427.1 | - | 2 | 119 | |||
FJ624078.1 | - | 2 | 47 | |||
FJ624079.1 | - | 2 | 119 | |||
V00518.1 | - | 3 | 169 |