Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144907.1 | NP_659156.1 | 4, 5 | 836 | ||
GenBank mRNA | AK143788.1 | - | 4, 5 | 880 | ||
AK170547.1 | - | 880 | ||||
AK182914.1 | - | 2, 3 | 256 | |||
AK196889.1 | - | 1, 2 | 65 | |||
BC005672.1 | - | 4, 5 | 836 | 3153129 |