Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144948.5 | NP_659197.2 | 2 | 321 | ||
GenBank mRNA | AF458961.1 | - | 2 | 285 | ||
AK005200.1 | - | 2 | 297 | |||
AK006755.1 | - | 2 | 286 | |||
AK008645.1 | - | 2 | 281 | |||
AK011223.1 | - | 2 | 303 | |||
AK015662.1 | - | 2 | 322 | |||
AK145381.1 | - | 2 | 285 | |||
AK151056.1 | - | 2 | 300 | |||
AK151827.1 | - | 2 | 316 | |||
AK153504.1 | - | 3 | 390 | |||
AK164653.1 | - | 1 | 1263 | |||
AK213555.1 | - | 1 | 138 | |||
AK216808.1 | - | 1 | 138 | |||
BC054774.1 | - | 2 | 288 | 6807637 | ||
BC116931.1 | - | 2 | 270 | 40126250 | ||
BC119038.1 | - | 2 | 270 | 8733787 | ||
BY087243.1 | - | 2 | 322 |