Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 3 | 649 | ||
NM_001099773.1 | NP_001093243.1 | 3 | 723 | |||
GenBank mRNA | AK056794.1 | - | 3 | 718 | ||
AK225715.1 | - | 3 | 555 | |||
AK292300.1 | - | 3 | 555 | |||
AK292301.1 | - | 2 | 513 | |||
AK310362.1 | - | 3 | 555 | |||
BC032329.1 | - | 3 | 538 | 5165263 | ||
BX398579.2 | - | 3 | 649 | |||
KU178011.1 | - | 3 | 494 | |||
M14565.1 | - | 3 | 538 |