Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145123.4 | NP_660105.3 | 6 | 975 | ||
GenBank mRNA | AJ421516.1 | - | 6 | 825 | ||
AK032328.1 | - | 6 | 840 | |||
AK049801.1 | - | 6 | 946 | |||
AK083221.1 | - | 6 | 979 | |||
AK149020.1 | - | 6 | 981 | |||
AK149069.1 | - | 6 | 976 | |||
BC024472.1 | - | 3 | 411 | 4976509 |