Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145152.4 | NP_660134.1 | 2 | 683 | ||
GenBank mRNA | AK043535.1 | - | 2 | 658 | ||
AY061858.1 | - | 2 | 656 | |||
BC058363.1 | - | 1 | 465 | |||
BC115652.2 | - | 1 | 491 | 40103158 |