Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145155.3 | NP_660137.1 | 4, 5 | 703 | ||
GenBank mRNA | AF454703.1 | - | 3, 4 | 322 | ||
AK044481.1 | - | 724 | ||||
BC027038.1 | - | 4, 5 | 703 | 5363785 |