Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144930.2 | NP_659179.1 | 14 | 1611 | ||
GenBank mRNA | AK136491.1 | - | 14 | 1603 | ||
BC013479.1 | - | 14 | 1586 | 4239756 |