Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144934.3 | NP_659183.3 | 3 | 1153 | ||
GenBank mRNA | AF503920.1 | - | 2 | 987 | ||
BC064744.1 | - | 3 | 1114 | 30051497 |