Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144942.4 | NP_659191.1 | 15 | 1797 | ||
GenBank mRNA | AK005015.1 | - | 15 | 1798 | ||
AY033912.1 | - | 15 | 1714 |