Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159375.1 | NP_001152847.1 | 6 | 709 | ||
NM_144958.4 | NP_659207.1 | 6 | 709 | |||
GenBank mRNA | AK010644.1 | - | 6 | 611 | ||
AK011529.1 | - | 6 | 602 | |||
AK077429.1 | - | 6 | 602 | |||
AK088124.1 | - | 4 | 1832 | |||
AK135752.1 | - | 6 | 602 | |||
AK135994.1 | - | 6 | 603 | |||
AK144850.1 | - | 6 | 602 | |||
AK145375.1 | - | 6 | 604 | |||
AK145508.1 | - | 6 | 604 | |||
AK146424.1 | - | 6 | 604 | |||
AK146464.1 | - | 6 | 602 | |||
AK152210.1 | - | 3 | 385 | |||
AK152694.1 | - | 6 | 604 | |||
AK159643.1 | - | 6 | 604 | |||
AK162017.1 | - | 6 | 604 | |||
AK166439.1 | - | 6 | 602 | |||
AK166527.1 | - | 6 | 602 | |||
AK166572.1 | - | 6 | 604 | |||
AK166674.1 | - | 6 | 604 | |||
AK167003.1 | - | 6 | 602 | |||
AK167083.1 | - | 6 | 602 | |||
AK167196.1 | - | 6 | 604 | |||
AK167205.1 | - | 6 | 601 | |||
AK167906.1 | - | 6 | 604 | |||
AK167988.1 | - | 6 | 602 | |||
AK168697.1 | - | 6 | 603 | |||
AK168780.1 | - | 6 | 602 | |||
AK169023.1 | - | 6 | 602 | |||
AK169030.1 | - | 6 | 595 | |||
AK169057.1 | - | 6 | 602 | |||
AK169156.1 | - | 6 | 602 | |||
AK169279.1 | - | 6 | 602 | |||
AK185730.1 | - | 2 | 86 | |||
AK187979.1 | - | 1 | 86 | |||
AK196145.1 | - | 86 | ||||
AK213871.1 | - | 3 | 260 | |||
AK215233.1 | - | 1 | 86 | |||
AK219842.1 | - | 1 | 86 | |||
BC012282.1 | - | 6 | 591 | |||
BC049915.1 | - | 6 | 592 | 5068709 | ||
BC099392.1 | - | 5 | 585 | |||
X03039.1 | - | 6 | 599 | |||
X03040.1 | - | 5 | 602 |