Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039545.2 | NP_001034634.2 | 25, 26 | 3390 | ||
GenBank mRNA | BC008538.1 | - | 24, 25 | 3329 | 3495412 |