Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145155.3 | NP_660137.1 | 7 | 1012 | ||
GenBank mRNA | AF454703.1 | - | 6 | 631 | ||
AK044481.1 | - | 1033 | ||||
BC027038.1 | - | 7 | 1012 | 5363785 |