Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145158.3 | NP_660140.1 | 4 | 1751 | ||
GenBank mRNA | AF468645.1 | - | 4 | 1702 | ||
AK154230.1 | - | 4 | 1721 | |||
AK155037.1 | - | 4 | 1731 | |||
AK155306.1 | - | 4 | 1745 | |||
AK155366.1 | - | 4 | 1731 | |||
AK155554.1 | - | 4 | 1747 | |||
AK155797.1 | - | 4 | 1752 | |||
AK155878.1 | - | 4 | 1749 | |||
AK155879.1 | - | 4 | 1745 | |||
AK155909.1 | - | 4 | 1751 | |||
AK155913.1 | - | 4 | 1721 | |||
AK155918.1 | - | 4 | 1778 | |||
AK170081.1 | - | 4 | 1748 | |||
AK170288.1 | - | 4 | 1721 | |||
AK170391.1 | - | 4 | 1721 | |||
AK170801.1 | - | 4 | 1752 | |||
AK170822.1 | - | 4 | 1773 | |||
AK170953.1 | - | 4 | 1767 | |||
AK171015.1 | - | 4 | 1773 | |||
AK171020.1 | - | 4 | 1739 | |||
AK171303.1 | - | 4 | 1723 | |||
AK171309.1 | - | 4 | 1748 | |||
AK171579.1 | - | 4 | 1721 | |||
AK171592.1 | - | 4 | 1721 | |||
AK171616.1 | - | 4 | 1721 | |||
BC053753.1 | - | 4 | 2035 | 6510220 | ||
GQ872457.1 | - | 4 | 1646 | |||
GQ872460.1 | - | 4 | 1646 |