Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145220.2 | NP_660255.1 | 4 | 323 | ||
GenBank mRNA | AK154525.1 | - | 4 | 347 | ||
AK160075.1 | - | 4 | 302 | |||
AK160105.1 | - | 4 | 299 | |||
AK161003.1 | - | 4 | 330 | |||
AK168403.1 | - | 4 | 284 | |||
AK168748.1 | - | 4 | 279 | |||
AK171512.1 | - | 4 | 279 | |||
AY113706.1 | - | 4 | 325 | |||
BC048906.1 | - | 4 | 261 | 6475576 |