Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145226.2 | NP_660261.1 | 2 | 420 | ||
GenBank mRNA | AB067534.1 | - | 2 | 420 | ||
AF453830.1 | - | 2 | 412 | |||
AK158392.1 | - | 2 | 402 | |||
BC141055.1 | - | 2 | 413 | 9055667 |