Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145227.3 | NP_660262.2 | 1 | 282 | ||
GenBank mRNA | AB067535.1 | - | 1 | 287 | ||
AF418010.1 | - | 1 | 222 | |||
BC118538.1 | - | 1 | 222 | |||
BU694306.1 | - | 1 | 282 |