Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 2 | 353 | ||
GenBank mRNA | AF197931.1 | - | 1 | 113 | ||
AK011822.1 | - | 2 | 340 | |||
AK042644.1 | - | 1 | 25 | |||
AK149957.1 | - | 2 | 186 | |||
BC011161.1 | - | 2 | 314 | |||
BC019384.1 | - | 2 | 334 | 5123736 |