Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290472.1 | NP_001277401.1 | 2 | 942 | ||
NM_001290473.1 | NP_001277402.1 | 2 | 942 | |||
NM_145355.5 | NP_663330.2 | 3 | 1061 | |||
GenBank mRNA | AK034108.1 | - | 3 | 431 | ||
AK052436.1 | - | 2 | 351 | |||
AK053358.1 | - | 2 | 308 | |||
AK075756.1 | - | 2 | 943 | |||
AK077638.1 | - | 2 | 273 | |||
AK149640.1 | - | 2 | 294 | |||
AK165895.1 | - | 367 | ||||
AK167455.1 | - | 2 | 259 | |||
AK168904.1 | - | 2 | 296 | |||
BC014812.1 | - | 2 | 273 | 3153870 |