Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 2 | 204 | ||
NM_001311312.1 | NP_001298241.1 | 2 | 179 | |||
XM_005264259.4 | XP_005264316.1 | 2 | 163 | |||
XM_011532764.2 | XP_011531066.1 | 2 | 163 | |||
GenBank mRNA | AK222827.1 | - | 2 | 164 | ||
AK225663.1 | - | 2 | 163 | |||
AK310483.1 | - | 2 | 143 | |||
AK313126.1 | - | 2 | 172 | |||
BC013979.2 | - | 2 | 126 | 4129593 | ||
BM704752.1 | - | 2 | 179 | |||
DC296316.1 | - | 2 | 205 | |||
EU847509.1 | - | 2 | 101 | |||
L17128.1 | - | 2 | 253 | |||
M81592.1 | - | 2 | 184 |