Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199676.1 | NP_001186605.1 | 3, 4 | 729 | ||
NM_145358.2 | NP_663333.1 | 3, 4 | 729 | |||
GenBank mRNA | AF453383.1 | - | 2, 3 | 512 | ||
AK031399.1 | - | 3, 4 | 654 | |||
AK032070.1 | - | 3, 4 | 702 | |||
AK044660.1 | - | 3, 4 | 718 | |||
AK160407.1 | - | 3, 4 | 645 | |||
BC023103.1 | - | 2, 3 | 562 | 3993474 |