Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012474.4 | NP_036606.2 | 4 | 776 | ||
XM_011509970.2 | XP_011508272.2 | 2 | 311 | |||
XM_017002236.1 | XP_016857725.1 | 5 | 981 | |||
GenBank mRNA | AB062451.1 | - | 4 | 395 | ||
AF236637.1 | - | 4 | 455 | |||
BC002906.2 | - | 4 | 616 | 3940564 |