Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145387.4 | NP_663362.2 | 4 | 913 | ||
GenBank mRNA | AK077662.1 | - | 4 | 914 | ||
AK085600.1 | - | 4 | 929 | |||
AK171694.1 | - | 3 | 823 | |||
AK194509.1 | - | 1 | 120 | |||
BC023151.1 | - | 3 | 599 | |||
BC056953.1 | - | 4 | 875 | 5717810 |