Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170556.1 | NP_001164027.1 | 1, 2 | 188 | ||
NM_145401.2 | NP_663376.2 | 5, 6 | 1270 | |||
GenBank mRNA | AK010703.1 | - | 1, 2 | 130 | ||
AK154296.1 | - | 5, 6 | 1271 | |||
AK165834.1 | - | 1, 2 | 130 | |||
AY348864.1 | - | 1, 2 | 121 | |||
BC015283.1 | - | 5, 6 | 1058 | 4238045 | ||
BQ572978.1 | - | 1, 2 | 121 |