Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145415.2 | NP_663390.2 | 1, 2 | 216 | ||
GenBank mRNA | AK052355.1 | - | 1, 2 | 217 | ||
AK088756.1 | - | 1, 2 | 217 | |||
AK147251.1 | - | 1, 2 | 210 | |||
BC019218.1 | - | 1, 2 | 197 | 5065857 |