Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145423.2 | NP_663398.2 | 1 | 626 | ||
GenBank mRNA | AK039927.1 | - | 1 | 455 | ||
AK136457.1 | - | 1 | 638 | |||
AY484428.1 | - | 1 | 626 | |||
BC017691.1 | - | 581 | 4239161 |