Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145425.3 | NP_663400.2 | 9 | 697 | ||
GenBank mRNA | AF424701.1 | - | 9 | 660 | ||
AK043672.1 | - | 8 | 584 | |||
AK082370.1 | - | 9 | 608 | |||
AK083042.1 | - | 9 | 698 | |||
BC056139.1 | - | 1 | 31 | |||
BC141166.1 | - | 9 | 661 | 9055778 |