Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145426.2 | NP_663401.2 | 2 | 429 | ||
NM_180599.1 | NP_850930.1 | 2 | 393 | |||
GenBank mRNA | AK042988.1 | - | 2 | 377 | ||
AK076071.1 | - | 2 | 395 | |||
AK080405.1 | - | 2 | 373 | |||
AK155122.1 | - | 2 | 407 | |||
BC006828.1 | - | 2 | 388 | 3597878 | ||
BC058567.1 | - | 1 | 337 | |||
CF174749.1 | - | 2 | 393 |