Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 6 | 495 | ||
GenBank mRNA | AK030445.1 | - | 6 | 375 | ||
AK033566.1 | - | 6 | 466 | |||
AK045438.1 | - | 6 | 431 | |||
AK049307.1 | - | 6 | 482 | |||
AK088080.1 | - | 6 | 446 | |||
AK146356.1 | - | 6 | 466 | |||
AY113705.1 | - | 6 | 460 | |||
BC063751.1 | - | 6 | 507 | 6511401 | ||
BU514763.1 | - | 6 | 506 |