Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145226.2 | NP_660261.1 | 4 | 894 | ||
GenBank mRNA | AB067534.1 | - | 4 | 894 | ||
AF453830.1 | - | 4 | 886 | |||
AK158392.1 | - | 4 | 876 | |||
BC141055.1 | - | 4 | 887 | 9055667 |