Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145374.2 | NP_663349.2 | 9 | 2367 | ||
GenBank mRNA | AK034623.1 | - | 1 | 1696 | ||
AK079844.1 | - | 4 | 292 | |||
BC020002.1 | - | 9 | 2365 | 3584335 | ||
BC025652.1 | - | 7 | 1435 |