Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042259.1 | - | 2 | 1445 | ||
BC019755.1 | - | 9, 10 | 1064 | 3964798 |