Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 5 | 462 | ||
NM_001102605.1 | NP_001096075.1 | 4 | 432 | |||
NM_001142573.1 | NP_001136045.1 | 2 | 232 | |||
NM_001142574.1 | NP_001136046.1 | 2 | 232 | |||
NM_001142575.1 | NP_001136047.1 | 2 | 232 | |||
NM_001142576.1 | NP_001136048.1 | 4 | 363 | |||
NM_001304521.1 | NP_001291450.1 | 2 | 260 | |||
NM_183243.2 | NP_899066.1 | 3 | 354 | |||
XM_017012168.1 | XP_016867657.1 | 4 | 432 | |||
XM_017012169.1 | XP_016867658.1 | 3 | 354 | |||
XM_017012170.1 | XP_016867659.1 | 2 | 261 | |||
XM_017012171.1 | XP_016867660.1 | 4 | 550 | |||
XM_017012172.1 | XP_016867661.1 | 4 | 550 | |||
XM_017012173.1 | XP_016867662.1 | 4 | 432 | |||
GenBank mRNA | AK054640.1 | - | 2 | 204 | ||
AK054667.1 | - | 3 | 354 | |||
AK092452.1 | - | 4 | 399 | |||
AK122994.1 | - | 4 | 431 | |||
AK131294.1 | - | 2 | 179 | |||
AK293413.1 | - | 2 | 204 | |||
BC033622.1 | - | 3 | 345 | 5210847 | ||
BX406207.2 | - | 3 | 357 | |||
CD014008.1 | - | 3 | 146 | |||
CD014010.1 | - | 4 | 246 | |||
CR933672.1 | - | 398 | ||||
DA980797.1 | - | 3 | 354 | |||
DC299597.1 | - | 2 | 232 | |||
DC326380.1 | - | 4 | 531 | |||
J05272.1 | - | 5 | 726 |