Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145422.4 | NP_663397.1 | 2 | 194 | ||
GenBank mRNA | AK145891.1 | - | 2 | 191 | ||
AK145916.1 | - | 2 | 191 | |||
AK149885.1 | - | 2 | 149 | |||
AK150376.1 | - | 2 | 153 | |||
AK150470.1 | - | 2 | 153 | |||
AK151042.1 | - | 2 | 187 | |||
AK151182.1 | - | 2 | 153 | |||
AK151932.1 | - | 2 | 153 | |||
AK152144.1 | - | 2 | 153 | |||
AK153434.1 | - | 2 | 153 | |||
AK167643.1 | - | 2 | 180 | |||
AK168038.1 | - | 2 | 188 | |||
AK172175.1 | - | 2 | 188 | |||
AY424364.1 | - | 2 | 187 | |||
BC003288.1 | - | 2 | 196 | 3583081 |