Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145423.2 | NP_663398.2 | 5 | 941 | ||
GenBank mRNA | AK039927.1 | - | 5 | 770 | ||
AK136457.1 | - | 5 | 951 | |||
AY484428.1 | - | 5 | 941 | |||
BC017691.1 | - | 896 | 4239161 |