Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145219.4 | NP_660254.1 | 8 | 2601 | ||
GenBank mRNA | AF515590.1 | - | 8 | 2590 | ||
AK047495.1 | - | 8 | 2552 | |||
AK049831.1 | - | 8 | 2554 | |||
AK087125.1 | - | 9 | 2740 | |||
AK132296.1 | - | 8 | 2595 | |||
BC055315.1 | - | 8 | 2461 | 5686778 | ||
BC061460.1 | - | 8 | 2461 | 5686778 |