Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 6 | 498 | ||
GenBank mRNA | AK030445.1 | - | 6 | 378 | ||
AK033566.1 | - | 6 | 469 | |||
AK045438.1 | - | 6 | 434 | |||
AK049307.1 | - | 6 | 485 | |||
AK088080.1 | - | 6 | 449 | |||
AK146356.1 | - | 6 | 469 | |||
AY113705.1 | - | 6 | 463 | |||
BC063751.1 | - | 6 | 510 | 6511401 | ||
BU514763.1 | - | 6 | 509 |