Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 9 | 1269 | ||
GenBank mRNA | AK011822.1 | - | 9 | 1256 | ||
AK042644.1 | - | 8 | 941 | |||
AK149957.1 | - | 9 | 1102 | |||
AK211539.1 | - | 1 | 18 | |||
AK215241.1 | - | 1 | 18 | |||
AK215883.1 | - | 1 | 18 | |||
AK219376.1 | - | 1 | 18 | |||
BC011161.1 | - | 9 | 1230 | |||
BC019384.1 | - | 9 | 1250 | 5123736 | ||
BC100575.1 | - | 8 | 1337 | 6432108 |