Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000888.4 | NP_000879.2 | 2 | 337 | ||
NM_001282353.1 | NP_001269282.1 | 3 | 230 | |||
NM_001282355.1 | NP_001269284.1 | 2 | 337 | |||
NM_001282389.1 | NP_001269318.1 | 2 | 337 | |||
GenBank mRNA | AK290300.1 | - | 2 | 305 | ||
AK313944.1 | - | 3 | 221 | |||
AL831998.1 | - | 2 | 276 | |||
BC121178.2 | - | 2 | 107 | 40122759 | ||
BC143656.1 | - | 2 | 115 | |||
CD013945.1 | - | 2 | 282 | |||
CD013947.1 | - | 2 | 282 | |||
DB222211.1 | - | 2 | 338 | |||
M35198.3 | - | 2 | 103 |