Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290472.1 | NP_001277401.1 | 7 | 3293 | ||
NM_001290473.1 | NP_001277402.1 | 6 | 3274 | |||
NM_145355.5 | NP_663330.2 | 8 | 3412 | |||
GenBank mRNA | AK034108.1 | - | 8 | 2782 | ||
AK052436.1 | - | 7 | 2702 | |||
AK053358.1 | - | 7 | 2659 | |||
AK077638.1 | - | 7 | 2624 | |||
AK149640.1 | - | 7 | 2644 | |||
AK165895.1 | - | 2715 | ||||
AK167455.1 | - | 7 | 2610 | |||
AK168904.1 | - | 7 | 2647 | |||
AV347010.1 | - | 133 | ||||
BC014812.1 | - | 7 | 2623 | 3153870 |