Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199676.1 | NP_001186605.1 | 9, 10 | 1120 | ||
NM_145358.2 | NP_663333.1 | 9, 10 | 1120 | |||
GenBank mRNA | AF453383.1 | - | 8, 9 | 903 | ||
AK031399.1 | - | 9, 10 | 1045 | |||
AK032070.1 | - | 9, 10 | 1093 | |||
AK044660.1 | - | 9, 10 | 1109 | |||
AK122370.1 | - | 4, 5 | 286 | |||
BC023103.1 | - | 8, 9 | 953 | 3993474 |