Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177874.1 | NP_001171345.1 | 4 | 493 | ||
NM_145370.2 | NP_663345.2 | 4 | 633 | |||
GenBank mRNA | AK088490.1 | - | 4 | 490 | ||
AK194760.1 | - | 1 | 97 | |||
BC003350.1 | - | 4 | 633 | 3481979 | ||
BC103782.1 | - | 4 | 460 | 6486599 |