Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145371.3 | NP_663346.1 | 7, 8 | 665 | ||
GenBank mRNA | AK088066.1 | - | 7, 8 | 664 | ||
AK133695.1 | - | 7, 8 | 666 | |||
BC003426.1 | - | 7, 8 | 665 | 2615801 |